2-Hydroxy-6-methylbenzonitrile ≥95%
Supplier: Matrix Scientific
074386-5G
102536-160EA
1053.85
USD
102536-160
2-Hydroxy-6-methylbenzonitrile ≥95%
2-Hydroxy-6-methylbenzonitrile
Hazard: Irritant
Formula:
C₈H₇NO |
MDL Number:
MFCD02261940 CAS Number: 73289-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...