4-Formyl-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methoxyisonicotinaldehyde2-Methoxypyridine-4-carbaldehyde

074383-1G
102536-144EA 72.31 USD
102536-144
4-Formyl-2-methoxypyridine ≥95%
4-Formyl-2-methoxypyridine

Hazard: Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD06410680
CAS Number: 72716-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR