3-Fluoro-5-iodobenzonitrile ≥95%
Supplier: Matrix Scientific
074382-1G
102536-140EA
1076.92
USD
102536-140
3-Fluoro-5-iodobenzonitrile ≥95%
3-Fluoro-5-iodobenzonitrile
Hazard: Irritant
Formula:
C₇H₃FIN MW: 247.0100 g/mol |
MDL Number:
MFCD04116468 CAS Number: 723294-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...