1,4-Dioxaspiro[4.5]decane-8-carbonitrile ≥95%

Supplier: Matrix Scientific

074373-1G
102536-090EA 181.54 USD
102536-090
1,4-Dioxaspiro[4.5]decane-8-carbonitrile ≥95%
1,4-Dioxaspiro[4.5]decane-8-carbonitrile

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.2050 g/mol
MDL Number: MFCD09037879
CAS Number: 69947-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR