1-(3-Methoxyphenyl)piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

074371-5G
102536-080EA 103.5 USD
102536-080
1-(3-Methoxyphenyl)piperazine dihydrochloride ≥95%
1-(3-Methoxyphenyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈Cl₂N₂O
MDL Number: MFCD00035285
CAS Number: 6968-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR