1-(4-Bromophenyl)piperazine hydrochloride ≥95%

Supplier: Matrix Scientific

074365-5G
102536-048EA 116.92 USD
102536-048
1-(4-Bromophenyl)piperazine hydrochloride ≥95%
1-(4-Bromophenyl)piperazine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄BrClN₂
CAS Number: 68104-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR