1-Bromo-5-chloro-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-chloro-1-nitrobenzene

074352-1G
102535-974EA 533.85 USD
102535-974
1-Bromo-5-chloro-2-nitrobenzene ≥95%
1-Bromo-5-chloro-2-nitrobenzene

Hazard: Irritant

Formula: C₆H₃BrClNO₂
MW: 236.4500 g/mol
MDL Number: MFCD08437614
CAS Number: 63860-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR