4-Boronobenzenesulfonamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Sulphamoylbenzeneboronic acid(4-Aminosulfonylphenyl)boronic acid(4-Aminosulphonylphenyl)boronic acid4-Sulfamoylbenzeneboronic acid

074336-5G
102535-894EA 606.15 USD
102535-894
4-Boronobenzenesulfonamide ≥95%
4-Boronobenzenesulphonamide

Hazard: Irritant/Store in Freezer

Formula: C₆H₈BNO₄S
MW: 201.00 g/mol
Melting Pt: 250…256 °C
MDL Number: MFCD06659839
CAS Number: 613660-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR