4-Fluoro-3-nitroanisole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-4-methoxy-2-nitrobenzene

074335-5G
102535-888EA 100 USD
102535-888
4-Fluoro-3-nitroanisole ≥95%
4-Fluoro-3-nitroanisole

Hazard: Irritant

Formula: C₇H₆FNO₃
MW: 171.13 g/mol
MDL Number: MFCD11848442
CAS Number: 61324-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR