5-Bromo-m-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-5-methylbenzoic acid

074318-1G
102535-800EA 29.9 USD
102535-800
5-Bromo-m-toluic acid ≥95%
5-Bromo-m-toluic acid

Hazard: Irritant

Formula: C₈H₇BrO₂
MW: 215.0440 g/mol
MDL Number: MFCD02684926
CAS Number: 58530-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR