5-Bromo-2-chlorobenzonitrile ≥95%

Supplier: Matrix Scientific

074312-5G
102535-770EA 29.23 USD
102535-770
5-Bromo-2-chlorobenzonitrile ≥95%
5-Bromo-2-chlorobenzonitrile

Hazard: Irritant

Formula: C₇H₃BrClN
MW: 216.46 g/mol
MDL Number: MFCD00672948
CAS Number: 57381-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR