3-Fluoro-2-nitroaniline ≥95%

Supplier: Matrix Scientific

074306-1G 074306-10G
102535-734EA 427.69 USD
102535-734 102535-738
3-Fluoro-2-nitroaniline ≥95%
3-Fluoro-2-nitroaniline

Hazard: Irritant

Formula: C₆H₅FN₂O₂
MW: 156.1145 g/mol
MDL Number: MFCD07368750
CAS Number: 567-63-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR