2-Hydroxy-6-methylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Cresotic acid6-Hydroxy-o-toluic acid6-Methylsalicylic acid

074305-5G
102535-730EA 227.69 USD
102535-730
2-Hydroxy-6-methylbenzoic acid ≥95%
2-Hydroxy-6-methylbenzoic acid

Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD01194284
CAS Number: 567-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR