1-(4-Hydroxyphenyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: p-(1-Piperazinyl)phenol4-(1-Piperazinyl)phenol4-(Piperazin-1-yl)phenol

074304-100G
102535-726EA 590.77 USD
102535-726
1-(4-Hydroxyphenyl)piperazine ≥95%
1-(4-Hydroxyphenyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
MDL Number: MFCD00066156
CAS Number: 56621-48-8
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR