2-Amino-4,6-dichlorophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichloro-6-hydroxyaniline

074286-100G
102535-634EA 156.92 USD
102535-634
2-Amino-4,6-dichlorophenol ≥95%
2-Amino-4,6-dichlorophenol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
MDL Number: MFCD00035766
CAS Number: 527-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR