4-Methoxypyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

074285-1G
102535-630EA 1076.92 USD
102535-630
4-Methoxypyridin-2(1H)-one ≥95%
4-Methoxypyridin-2(1H)-one

Hazard: Irritant

Formula: C₆H₇NO₂
MW: 125.1253 g/mol
MDL Number: MFCD09839191
CAS Number: 52545-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR