2,6-Dimethyl-2,5-heptadien-4-one ≥95%

Supplier: Matrix Scientific

074268-5G
102535-538EA 58.46 USD
102535-538
2,6-Dimethyl-2,5-heptadien-4-one ≥95%
2,6-Dimethyl-2,5-heptadien-4-one

Hazard: Irritant

Formula: C₉H₁₄O
MW: 138.2069 g/mol
Boiling Pt: 198…199 °C
Melting Pt: 23…26 °C
MDL Number: MFCD00008901
CAS Number: 504-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR