2,5-Dichloro-4-aminophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2,5-dichlorophenol

074266-5G
102535-526EA 458.46 USD
102535-526
2,5-Dichloro-4-aminophenol ≥95%
2,5-Dichloro-4-aminophenol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MW: 178.0200 g/mol
MDL Number: MFCD09743861
CAS Number: 50392-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR