Ethyl-3-amino-4-fluorobenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-amino-4-fluorobenzoate
074247-10G
074247-1G
102535-424EA
213.85
USD
102535-424
102535-428
Ethyl-3-amino-4-fluorobenzoate ≥95%
Ethyl-3-amino-4-fluorobenzoate
Hazard: Irritant
Formula:
C₉H₁₀FNO₂ MW: 183.1796 g/mol |
MDL Number:
MFCD03840487 CAS Number: 455-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...