2-Fluoro-3-iodobenzoic acid ≥95%

Supplier: Matrix Scientific

074245-1G
102535-416EA 478.4 USD
102535-416
2-Fluoro-3-iodobenzoic acid ≥95%
2-Fluoro-3-iodobenzoic acid

Hazard: Irritant

Formula: C₇H₄FIO₂
MW: 266.0083 g/mol
MDL Number: MFCD07774248
CAS Number: 447464-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR