1-Boc-4-(4-bromobenzoyl)piperidine ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-4-(4-Bromobenzoyl)piperidine
074243-5G
074243-250MG
102535-404EA
355.38
USD
102535-404
102535-408
1-Boc-4-(4-bromobenzoyl)piperidine ≥95%
1-Boc-4-(4-bromobenzoyl)piperidine
Hazard: Irritant
Formula:
C₁₇H₂₂BrNO₃ MW: 368.2700 g/mol |
MDL Number:
MFCD04114991 CAS Number: 439811-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...