2,5-Diaminopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: Pyridine-2,5-diaminePyridine-2,5-diyldiamine

074239-1G
102535-384EA 284.62 USD
102535-384
2,5-Diaminopyridine ≥95%
2,5-Diaminopyridine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD00129023
CAS Number: 4318-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR