3-Methylmorpholine ≥95%
Supplier: Matrix Scientific
074238-1G
102535-380EA
769.23
USD
102535-380
3-Methylmorpholine ≥95%
3-Methylmorpholine
Hazard: Irritant
Formula:
C₅H₁₁NO MW: 101.1469 g/mol |
MDL Number:
MFCD09054773 CAS Number: 42185-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...