4-Fluoro-3-iodobenzoic acid ≥95%

Supplier: Matrix Scientific

074233-5G
102535-352EA 284.05 USD
102535-352
4-Fluoro-3-iodobenzoic acid ≥95%
4-Fluoro-3-iodobenzoic acid

Hazard: Irritant

Formula: C₇H₄FIO₂
MW: 266.01 g/mol
MDL Number: MFCD03788556
CAS Number: 403-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR