1-Bromo-2-chloro-3-nitrobenzene ≥95%
Supplier: Matrix Scientific
074230-5G
102535-336EA
315.38
USD
102535-336
1-Bromo-2-chloro-3-nitrobenzene ≥95%
1-Bromo-2-chloro-3-nitrobenzene
Hazard: Irritant
Formula:
C₆H₃BrClNO₂ MW: 236.4505 g/mol |
MDL Number:
MFCD08277301 CAS Number: 3970-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...