4-N-Fmoc-aminocyclohexanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-N-Fmoc-Amino-cyclohexanone

074226-5G
102535-312EA 921.54 USD
102535-312
4-N-Fmoc-aminocyclohexanone ≥95%
4-N-Fmoc-aminocyclohexanone

Hazard: Irritant

Formula: C₂₁H₂₁NO₃
MW: 335.4000 g/mol
MDL Number: MFCD03001741
CAS Number: 391248-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR