2-Acetyl-6-methoxynaphthalene ≥95%
Supplier: Matrix Scientific
Synonyms:
6'-Methoxy-2'-acetonaphthone
074225-10G
074225-100G
074225-5G
102535-304EA
15.38
USD
102535-304
102535-308
102535-306
2-Acetyl-6-methoxynaphthalene ≥95%
2-Acetyl-6-methoxynaphthalene
Hazard: Irritant
Formula:
C₁₃H₁₂O₂ MW: 200.24 g/mol |
MDL Number:
MFCD00021643 CAS Number: 3900-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...