2-Phosphonobutane-1,2,4-tricarboxylic acid 50% in aqueous solution , 95%

Supplier: Matrix Scientific

Synonyms: PBTC3-Carboxy-3-phosphonohexanedioic acid

074215-100G
102535-254EA 153.85 USD
102535-254
2-Phosphonobutane-1,2,4-tricarboxylic acid 50% in aqueous solution , 95%
2-Phosphonobutane-1,2,4-tricarboxylic acid

Hazard: Corrosive

Formula: C₇H₁₁O₉P
MDL Number: MFCD01940753
CAS Number: 37971-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR