Methyl-8-methyl-3-oxo-8-azabicyclo[3.2.1]octane-2-carboxylate ≥95%

Supplier: Matrix Scientific

074206-1G 074206-5G
102535-206EA 283.08 USD
102535-206 102535-208
Methyl-8-methyl-3-oxo-8-azabicyclo[3.2.1]octane-2-carboxylate ≥95%
Methyl-8-methyl-3-oxo-8-azabicyclo[3.2.1]octane-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
CAS Number: 36127-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR