2-Acetyl-4-cyanophenol ≥95%

Supplier: Matrix Scientific

074205-5G 074205-1G
102535-204EA 1000 USD
102535-204 102535-202
2-Acetyl-4-cyanophenol ≥95%
2-Acetyl-4-cyanophenol

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.1600 g/mol
MDL Number: MFCD00100432
CAS Number: 35794-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR