5,6,7,8-Tetrahydroquinoxaline ≥95%
Supplier: Matrix Scientific
074196-10G
074196-100G
102535-154EA
200
USD
102535-154
102535-152
5,6,7,8-Tetrahydroquinoxaline ≥95%
5,6,7,8-Tetrahydroquinoxaline
Hazard: Irritant
Formula:
C₈H₁₀N₂ MW: 134.18 g/mol |
MDL Number:
MFCD00038078 CAS Number: 34413-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...