(S)-(+)-3-Aminopiperidine dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-(+)-3-Piperidinamine dihydrochloride
, (S)-Piperidin-3-amine dihydrochloride
074191-5G
074191-10G
074191-1G
102535-128EA
769.23
USD
102535-128
102535-126
102535-124
(S)-(+)-3-Aminopiperidine dihydrochloride ≥95%
(S)-(+)-3-Aminopiperidine dihydrochloride
Hazard: Irritant
Formula:
C₅H₁₄Cl₂N₂ MW: 173.08 g/mol |
MDL Number:
MFCD03427036 CAS Number: 334618-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...