4-Nitrophthalonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-1,2-benzenedicarbonitrile4-Nitrobenzene-1,2-dicarbonitrile4-Nitro-1,2-dicyanobenzene

074180-5G
102535-068EA 16.92 USD
102535-068
4-Nitrophthalonitrile ≥95%
4-Nitrophthalonitrile

Hazard: Irritant

Formula: C₈H₃N₃O₂
MW: 173.13 g/mol
Melting Pt: 136…137 °C
MDL Number: MFCD00040301
CAS Number: 31643-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR