4-Hydroxy-7-methoxyquinoline-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

074165-5G 074165-1G 074165-10G
102534-990EA 880 USD
102534-990 102534-986 102534-988
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid ≥95%
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₄
MW: 219.1900 g/mol
MDL Number: MFCD00024004
CAS Number: 28027-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR