2-Bromo-4-iodobenzaldehyde ≥95%

Supplier: Matrix Scientific

074160-250MG 074160-1G 074160-5G
102534-958EA 400 USD
102534-958 102534-960 102534-962
2-Bromo-4-iodobenzaldehyde ≥95%
2-Bromo-4-iodobenzaldehyde

Hazard: Irritant

Formula: C₇H₄BrIO
MW: 310.9200 g/mol
MDL Number: MFCD07779025
CAS Number: 261903-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR