5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid5-Formyl-2,4-dimethyl-3-pyrrolecarboxylic acid

074154-5G
102534-926EA 153.85 USD
102534-926
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid ≥95%
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD06202342
CAS Number: 253870-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR