4,4-Difluorocyclohexanol ≥95%

Supplier: Matrix Scientific

074140-5G 074140-1G
102534-854EA 307.69 USD
102534-854 102534-856
4,4-Difluorocyclohexanol ≥95%
4,4-Difluorocyclohexanol

Hazard: Irritant/Refrigerate

Formula: C₆H₁₀F₂O
MW: 136.1398 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD10000566
CAS Number: 22419-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR