4-Hydroxy-3,5-dimethylbenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dimethyl-4-hydroxybenzaldehyde

074138-100G
102534-844EA 304.62 USD
102534-844
4-Hydroxy-3,5-dimethylbenzaldehyde ≥95%
4-Hydroxy-3,5-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00006946
CAS Number: 2233-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR