(R)-(+)-4-Methoxy-α-methylbenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(+)-1-(4-Methoxyphenyl)ethylamine(R)-1-(4-Methoxyphenyl)ethan-1-amine

074134-100G 074134-25G
102551-872EA 253.85 USD
102551-872 102534-822
(R)-(+)-4-Methoxy-α-methylbenzylamine ≥95%
(R)-(+)-4-Methoxy-α-methylbenzylamine

Hazard: Irritant

Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00671659
CAS Number: 22038-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR