5-Bromo-1,2,3-trichlorobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3,4,5-trichlorobenzene

074129-100G 074129-25G 074129-5G
102551-870EA 304.62 USD
102551-870 102534-790 102833-672
5-Bromo-1,2,3-trichlorobenzene ≥95%
5-Bromo-1,2,3-trichlorobenzene

Hazard: Irritant

Formula: C₆H₂BrCl₃
MW: 260.34 g/mol
MDL Number: MFCD00155009
CAS Number: 21928-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR