4-Bromo-1-methylpyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

074122-1G 074122-5G
102534-754EA 89.23 USD
102534-754 102534-756
4-Bromo-1-methylpyridin-2(1H)-one ≥95%
4-Bromo-1-methylpyridin-2(1H)-one

Hazard: Irritant

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Melting Pt: 107…110 °C
MDL Number: MFCD11226870
CAS Number: 214342-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR