7-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

074103-250MG 074103-1G
102534-656EA 826.15 USD
102534-656 102534-658
7-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%
7-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MW: 199.6800 g/mol
MDL Number: MFCD09026781
CAS Number: 19500-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR