6-Bromoisobenzofuran-1(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromophthalide

074102-5G 074102-1G
102534-650EA 90.77 USD
102534-650 102534-652
6-Bromoisobenzofuran-1(3H)-one ≥95%
6-Bromoisobenzofuran-1(3H)-one

Hazard: Irritant

Formula: C₈H₅BrO₂
MW: 213.0281 g/mol
MDL Number: MFCD03428561
CAS Number: 19477-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR