4-(2-Pyridyl)aniline ≥95%
Supplier: Matrix Scientific
074092-5G
074092-10G
074092-1G
102534-594EA
40
USD
102534-594
102534-596
102534-598
4-(2-Pyridyl)aniline ≥95%
4-(2-Pyridyl)aniline
Hazard: Irritant
Formula:
C₁₁H₁₀N₂ Melting Pt: 95…97 °C |
CAS Number:
18471-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...