(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Nα-Z-Nβ-Boc-L-2,3-diaminopropionic acidZ-Dap(Boc)-OH2-Benzyloxy-carbonyl-amino-3-tert-butoxycarbonyl-amino-propanoic-acidZ-3-(Boc-amino)-L-alanine

074072-5G
102534-494EA 200 USD
102534-494
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥95%
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₆
MW: 338.40 g/mol
MDL Number: MFCD00237342
CAS Number: 16947-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR