(S)-Piperazine-2-carboxylic acid dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (2S)-piperazine-2-carboxylic acid dihydrochloride

074055-1G 074055-5G 074055-10G
102534-396EA 46.15 USD
102534-396 102534-398 102534-400
(S)-Piperazine-2-carboxylic acid dihydrochloride ≥95%
(S)-Piperazine-2-carboxylic acid dihydrochloride

Hazard: Irritant

Formula: C₅H₁₂Cl₂N₂O₂
MW: 203.07 g/mol
MDL Number: MFCD01632104
CAS Number: 158663-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR