2,8-Quinolinediol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,8-Dihydroxyquinoline

074051-100G
102534-376EA 227.69 USD
102534-376
2,8-Quinolinediol ≥95%
2,8-Quinolinediol

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00216696
CAS Number: 15450-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR