1-(2,6-Dichlorophenyl)oxindole ≥95%

Supplier: Matrix Scientific

Synonyms: Diclofenac Amide1-(2,6-Dichlorophenyl)-2-indolinone1-(2,6-Dichlorophenyl)-2-oxoindoline

074049-5G
102534-364EA 53.85 USD
102534-364
1-(2,6-Dichlorophenyl)oxindole ≥95%
1-(2,6-Dichlorophenyl)oxindole

Hazard: Irritant

Formula: C₁₄H₉Cl₂NO
MW: 278.13 g/mol
MDL Number: MFCD00451393
CAS Number: 15362-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR