2,3-Dihydro-1H-carbazol-4(9H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
1,2,3,4-Tetrahydro-4-oxocarbazole
, 1,2,3,4-Tetrahydrocarbazol-4-one
074044-100G
074044-10G
102534-336EA
107.69
USD
102534-336
102534-338
2,3-Dihydro-1H-carbazol-4(9H)-one ≥95%
2,3-Dihydro-1H-carbazol-4(9H)-one
Hazard: Irritant
Formula:
C₁₂H₁₁NO MW: 185.2218 g/mol |
MDL Number:
MFCD00173749 CAS Number: 15128-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...