4-(4-Aminophenyl)butanoic acid ≥95%

Supplier: Matrix Scientific

074042-5G
102534-326EA 206.15 USD
102534-326
4-(4-Aminophenyl)butanoic acid ≥95%
4-(4-Aminophenyl)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MW: 179.2200 g/mol
MDL Number: MFCD00075052
CAS Number: 15118-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR